Learn More
Nip1 Mouse, Unlabeled, Clone: 5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611096
Description
Apoptosis, a selective process of genetically programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. In addition, apoptotic programs in virus-infected cells regulate viral replication and pathogenesis. However, viruses, such as human adenovirus, have evolved methods of circumventing these programs. The adenovirus E1B region encodes a 19kDa protein (19K) that, similar to Bcl-2, suppresses apoptosis via interactions with intracellular proteins such as Bcl-2 family members. In addition, 19K and Bcl-2 interact with members of a novel subfamily of pro-apoptotic proteins which includes Nip1-3 and Nix. These proteins contain C-terminal transmembrane (TM) domains, such as those found in Bcl-2 proteins. The TM domain is important for targeting of proteins to mitochondria and nuclear envelope/ER regions of the cell. More specifically, Nip1 localizes at the nuclear envelope/ER, while Nip3 localizes in mitochondria and other cytoplasmic membrane structures. In addition, Nip1 shares homology with the catalytic domain of mammalian calcium/calmodulin-dependent cyclic nucleotide phosphodiesterases (PDEs) and may have a PDE-like activity.
Immunofluorescence, Western Blotting
Specifications
Nip1 | |
Monoclonal | |
250μg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG1 |
Immunofluorescence, Western Blot | |
5 | |
Unconjugated | |
Human Nip1 aa. 52-174 | |
50 μg | |
Cell Biology | |
Human, Rat |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.