Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1038

Catalog No. PA1038


Add to cart

Description

Description

PA1-038 detects recombit rat brain nitric oxide synthase (nNOS) overexpressed in E. coli. This antibody does not to detect iNOS or eNOS. PA1-038 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~155 kDa protein representing recombit rat nNOS protein overexpressed in E. coli. The PA1-038 immunogen is a synthetic peptide corresponding to residues I(237) Q V D R D L D G K S H K A(250) of rat nNOS.

Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) & endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains. bNOS and eNOS share approximately 50% sequence homology and their enzymatic activity depends on binding to the calcium/calmodulin complex. Increases in intracellular calcium lead to the production of low levels of NO over a short time. bNOS is found in neurons, peripheral nerve cells, macula densa, and pancreatic islet cells. Alternate splicing specifically regulates bNOS in striated muscle. The translated protein, bNOS mu, is 34 amino acid residues larger than bNOS from brain.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

nNOS
Polyclonal
PBS with 1mg/mL BSA and 0.05% sodium azide
P29476
Neuronal Nitric Oxide Synthase, NOS Type 1, Brain NOS, bNOS1, bNOS
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24598
Western Blot
Unconjugated
Nos1
Liquid
NOS1
Synthetic peptide corresponding to residues I(237) Q V D R D L D G K S H K A(250) of rat nNOS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit