Learn More
anti-NKG2A/CD159a, Alexa Fluor 488, Clone: 705829, R&D Systems™
Manufacturer:  R&D Systems FAB6867G100
 View more versions of this product
Catalog No. FAB6867G100
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
NKG2A/CD159a/KLRC1
705829
Flow Cytometry 5 uL/10^6 cells
Q9Z202
CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Rat
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse NKG2A/CD159a in direct ELISAs.
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
KLRC1
CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a, Accession # Q9Z202
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3821
Mouse