Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-NKG2A/CD159a, APC, Clone: 705829, R&D Systems™
Specifications
Antigen | NKG2A/CD159a/KLRC1 |
---|---|
Clone | 705829 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB6867A025
|
R&D Systems
FAB6867A025 |
25 Tests | N/A | N/A | N/A | ||||
FAB6867A100
|
R&D Systems
FAB6867A100 |
100 Tests | N/A | N/A | N/A | ||||
Description
Applications:
Flow Cytometry
Specifications
NKG2A/CD159a/KLRC1 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
KLRC1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse NKG2A/CD159a in direct ELISAs. |
705829 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
Q9Z202 | |
3821 | |
CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a Accession # Q9Z202 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title