Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-NKG2C/CD159c, APC, Clone: 134591, R&D Systems
Manufacturer: R&D Systems FAB138A025
Description
Applications:
Flow Cytometry
Specifications
NKG2C/CD159c/KLRC2 | |
134591 | |
Flow Cytometry 10 uL/10^6 cells | |
Purified | |
KLRC2 | |
BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
3822 | |
Human |
Flow Cytometry | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
Mouse | |
IgG1 | |
25 Tests | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
Monoclonal | |
Detects human NKG2C/CD159c heterodimer with CD94 in flow cytometry. It does not cross-react with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title