Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIAMRE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14173

Catalog No. PA14173


Add to cart

Description

Description

PA1-4173 detects NKIAMRE in human samples. PA1-4173 has been successfully used in Western blot procedures. The PA1-4173 immunogen is a synthetic peptide corresponding to residues S(14) Y G T V M K C K H K N T G Q I(29) of human NKIAMRE.

A novel cyclin-dependent kinase-related molecule, NKIAMRE, has been identified that is thought to be a tumor suppressor locus. The NKIAMRE gene has a cytolocalization on 5q, which is a region that is interstitially deleted in human acute leukemia and human myeloid dysplastic disorders.
Specifications

Specifications

NKIAMRE
Polyclonal
Unconjugated
CDKL3
Liquid
CDKL3
Synthetic peptide corresponding to residues S(14) Y G T V M K C K H K N T G Q I(29) of human NKIAMRE.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8IVW4
Cyclin-dependent kinase-like 3, Serine/threonine protein kinase NKIAMRE, CDKL3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51265
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit