Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIAMRE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527912

Catalog No. PIPA527912


Add to cart

Description

Description

PA5-27912 targets NKIAMRE in WB applications and shows reactivity with Human samples. The PA5-27912 immunogen is recombit fragment corresponding to a region within amino acids 46 and 308 of Human NKIAMRE. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determit of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

NKIAMRE
Polyclonal
Unconjugated
CDKL3
Liquid
CDKL3
Recombit fragment corresponding to a region within amino acids 46 and 308 of Human NKIAMRE
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1.03 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q8IVW4
NKIAMRE, serine-threonine protein kinase NKIAMRE, serine/threonine-protein kinase NKIAMRE
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51265
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit