Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NKp46/NCR1 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
$149.64 - $428.04
Specifications
Antigen | NKp46/NCR1 |
---|---|
Clone | 195314 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1850P025
|
R&D Systems
FAB1850P025 |
25 Tests |
Each for $149.64
|
|
|||||
FAB1850P100
|
R&D Systems
FAB1850P100 |
100 Tests |
Each for $428.04
|
|
|||||
Description
NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry.Specifications
NKp46/NCR1 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
9437 | |
Mouse T cell hybridoma transfected with human NKp46/NCR1 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
195314 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
NCR1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human NKp46/NCR1 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title