Learn More
anti-NKp46/NCR1, PE, Clone: 195314, R&D Systems™
Manufacturer:  R&D Systems FAB1850P025
 View more versions of this product
Catalog No. FAB1850P025
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
NKp46/NCR1
195314
Flow Cytometry 10 uL/10^6 cells
Purified
NCR1
Mouse T cell hybridoma transfected with human NKp46/NCR1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9437
Human
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Mouse
IgG2b
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human NKp46/NCR1 in direct ELISAs and Western blots.