Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx2.2 Mouse, Alexa Fluor 647, Clone: 74.5A5, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564729

Catalog No. BDB564729


Add to cart

Description

Description

The 74.5A5 monoclonal antibody specifically binds to Homeobox protein Nkx-2.2 (Nkx2.2), which is a transcription factor required for the differentiation of ventral neuronal populations in the central nervous system and endocrine cell populations in the pancreas and intestine. Within the pancreas, Nkx2.2 is expressed in many cell subsets and plays important roles in islet and β- cell differentiation and maturation. Mice with a homozygous disruption of the Nkx2-2 gene lack mature β cells and die shortly after birth with severe hyperglycemia. Similarly to the Nkx2.2-ablated mice, homozygous mutations in the human NKX2-2 gene have been described in diabetic patients and associated with the etiology of neonatal diabetes. The NKX2-2 gene is also a target of the Ewing sarcoma specific protein EWS-FLI. Nkx2.2 expression can be monitored in order to determine cell fate changes in cultures for the generation of human pluripotent stem cell-derived insulin-producing cells as well as oligodendrocyte progenitors.

Specifications

Specifications

Nkx2.2
74.5A5
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
NKX2-2; NKX2B; Nkx2-2; Nkx-2.2; NK-2 homolog B; glycoprotein GP330
Affinity Purified
Chicken Nkx2.2 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Chicken, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit