Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.5 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP151953

Catalog No. NBP151953


Add to cart

Description

Description

NKX2.5 Polyclonal antibody specifically detects NKX2.5 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications

Specifications

NKX2.5
0.5 mg/mL
Western Blot 0.1 - 0.5 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000
NP_004378.1
cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2EFLJ97197, NKX4-1, tinman paralog
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize all three reported isoforms (NP_004378.1; NP_001159647.1; NP_001159648.1).
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NKX2-5
Peptide with sequence C-PRAYSDPDPAKDPR corresponding to internal region according to NP_001159647.1, NP_001159648.1.
Immunogen affinity purified
RUO
Primary
1482
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only