Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx2.8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527456

Catalog No. PIPA527456


Add to cart

Description

Description

PA5-27456 targets NKX2-8 in WB applications and shows reactivity with Human samples. The PA5-27456 immunogen is recombit fragment corresponding to a region within amino acids 1 and 170 of Human NKX2-8. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Nkx2.8
Polyclonal
Unconjugated
NKX2-8
Liquid
NKX2-8
Recombit fragment corresponding to a region within amino acids 1 and 170 of Human NKX2-8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O15522
NKX2.8, NKX2H, Nkx2-9, NK-2 homolog 8, NK-2 homolog H, NK2 transcription factor related, locus 8, homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26257
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit