Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nkx3.1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14395

Catalog No. PA14395


Add to cart

Description

Description

PA1-4395 detects NKX3.1 from human samples. PA1-4395 has been successfully used in Western blot procedures. PA1-4395 immunizing peptide corresponds to amino acid residues 21-37 from human NKX3.1 protein.

NKX3.1 is a homeobox gene which exhibits prostate and testis specific expression. Loss of NKX3.1 expression has been implicated in prostate development and tumorigenesis. NKX3.1 expression is dramatically down-regulated in testicular cancer of germ cell origin. Results suggest that NKX3.1 is essential for normal testis function and that its loss of expression is highly associated with the invasive phenotype of testicular germ cell tumors.
Specifications

Specifications

NKX3.1
Polyclonal
Unconjugated
NKX3-1
Liquid
NKX3-1
Synthetic Peptide: T(21) P S K P L T S F L I Q D I L R D(37)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q99801
Homeobox protein Nkx-3.1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4824
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit