Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NLRX1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP149838

Catalog No. NBP149838

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

NLRX1
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
NP_078894.2
Caterpiller protein 11.3, CLR11.3DLNB26, MGC131937, MGC21025, NLR family member X1, NLR family, X1, NOD26, NOD5, NOD9FLJ21478, NOD-like receptor X1, Nucleotide-binding oligomerization domain protein 26, Nucleotide-binding oligomerization domain protein 5, Nucleotide-binding oligomerization domain protein 9, nucleotide-binding oligomerization domain, leucine rich repeat containing X1
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_078894.2; NP_733840.1).
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
NLRX1
Peptide with sequence C-NFSGETLDSTDPSN corresponding to internal region according to NP_078894.2, NP_733840.1.
Immunogen affinity purified
RUO
Primary
79671
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only