Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nm23 Mouse, Unlabeled, Clone: 56, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610247

 View more versions of this product

Catalog No. BDB610247


Add to cart

Description

Description

Nm23 is a potential metastasis suppressor protein whose expression is either reduced, altered (by mutation), or amplified in various types of metastatic carcinomas. Two human gene homologues, nm23-H1 and nm23-H2, encode 17 kDa proteins that are 90% identical. Nm23 proteins possess a nucleoside diphosphate kinase (NDPK) activity. This enzymatic activity catalyzes the synthesis of non-adenine-containing nucleoside triphosphates from nucleoside diphosphates via a phosphorylated enzyme intermediate. In addition, Nm23 inhibits differentiation, interacts with GTP-binding (GAP) proteins, autophosphorylates serine residues, and binds to DNA. The biochemical mechanisms by which Nm23 affects tumor metastatic potential have yet to be determined. In murine melanoma cell lines, serine 44 is the major site of autophosphorylation on Nm23-1. This acid-stable phosphorylation of Nm23 is inhibited in vitro by cAMP and in vivo by forskolin. These data indicate that this serine phosphorylation is regulated via some cAMP-dependent event in signal transduction. In addition, it has been shown that the Nm23-H2 protein is identical to the c-myc transcription factor PuF. This suggests that some of the cellular effects of Nm23 are mediated by its transcriptional regulatory function, while others are mediated by its NDPK activity.

Specifications

Specifications

Nm23
56
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Chicken, Human, Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.