Learn More
NMB Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525267
Catalog No. PIPA525267
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with mouse based on sequence homology.
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.
Specifications
Specifications
NMB
Polyclonal
Unconjugated
NMB
Liquid
NMB
KLH conjugated synthetic peptide between 15-42 amino acids from the central region of human NMB
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P08949
neuromedin-B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4828