Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA34664
Description
This antibody is specific for the ∽180 kDa NR2A subunit of the NMDA Receptor. There is no reactivity towards the NR2B and NR2C subunits. Immunolabeling is blocked by preadsorption of antibody with the fusion protein used to generate the antibody.
NMDAR2A Polyclonal antibody specifically detects NMDAR2A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NMDAR2A | |
Polyclonal | |
Unconjugated | |
Grin2a | |
EPND; FESD; GluN2A; GluRepsilon1; glutamate [NMDA] receptor subunit epsilon-1; glutamate ionotropic receptor NMDA type subunit 2A; glutamate receptor; glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, NMDA2A (epsilon 1); glutamate receptor, ionotropic, N-methyl D-aspartate 2A; Grin2a; hNR2A; LKS; NMDA Receptor 2A; NMDAR 2A; NMDAR2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NR2A; OTTHUMP00000160135; OTTHUMP00000174531 | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
P35436, Q00959, Q12879 | |
Grin2a | |
Fusion protein from the C-terminal region of the NR2A subunit of rat GRIN2A | |
RUO | |
14811, 24409, 2903 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction