Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527921

Catalog No. PIPA527921


Add to cart

Description

Description

PA5-27921 targets NMDAR2A in IHC (P) and WB applications and shows reactivity with Human, Rat and mouse samples. The PA5-27921 immunogen is recombit fragment corresponding to a region within amino acids 173 and 510 of Human NMDAR2A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

NMDAR2A
Polyclonal
Unconjugated
GRIN2A
Liquid
GRIN2A
Recombit fragment corresponding to a region within amino acids 173 and 510 of Human NMDAR2A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q12879, P35436
EPND, GluN2A, NMDAR2A, NR2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor channel, subunit epsilon-1, N-methyl-D-aspartate receptor subunit 2A, NMDA receptor subtype 2A, glutamate [NMDA] receptor subunit epsilon-1, hNR2A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14811, 2903
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit