Learn More
anti-NMDAR2D, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP146482
Catalog No. NBP146482
Add to cart
Description
Description
NMDAR2D Polyclonal antibody specifically detects NMDAR2D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
NMDAR2D
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Antisera
GRIN2D
A synthetic peptide from 2d cytoplasmic domain of human NMDAR2D (GRIN2D) conjugated to an immunogenic carrier protein was used as the antigen.
Unpurified
RUO
Primary
2906
Reconstitute with 0.1 ml sterilized water to desired concentration.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
EB11estrogen receptor binding CpG island, glutamate receptor, ionotropic, N-methyl D-aspartate 2D, NMDA epsilon 4, NMDAR2DN-methyl-d-aspartate receptor subunit 2D, N-methyl D-aspartate receptor subtype 2D, NR2Dglutamate [NMDA] receptor subunit epsilon-4
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specific for NMDAR2D.
Human