Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NME1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127872

Catalog No. PIPA127872

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Major role in the synthesis of nucleoside triphosphates other than ATP. This protein (NME1) was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (e

PA1-27872 detects NME1 from human samples.PA1-27872 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on HEK 293 cell lysate.The immunogen is a synthetic peptide corresponding to human Nucleoside diphosphate kinase A.
Specifications

Specifications

NME1
Polyclonal
PBS with no preservative
P15531
AWD, GAAD, NB, NBS, NDPK-A, NDPKA, NM23, NM23-H1
Chicken
IgY
50μg
Store at -20°C
Primary
4830
Western Blot
Unconjugated
NME1
Purified
NME1
Synthetic peptide corresponding to human Nucleoside diphosphate kinase A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit