Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS/NOS Type I Rabbit, Unlabeled, Polyclonal, BD

Rabbit Polyclonal Antibody

Manufacturer:  BD Biosciences 610310

 View more versions of this product

Catalog No. BDB610310


Add to cart

Description

Description

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+ levels and enhance calmodulin binding. Neuronal NOS (nNOS or bNOS) and endothelial NOS (ECNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin and are regulated in a similar manner. However, both have been shown to be distinct gene products of about 155kDa and 140kDa, respectively, and the human forms show 52% amino acid identity. Neuronal NOS and induced macrophage NOS (iNOS) share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain. Neuronal NOS, a cytosolic protein present mainly in neural tissues, has been purified and characterized from rat cerebellum. The NO synthesized by this enzyme acts as a neurotransmitter. ECNOS has been cloned from human vascular endothelium as well as from bovine aortic endothelial cells (BAEC) and has a unique N-myristylation consensus sequence that may explain its membrane localization.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

nNOS/NOS Type I
POLYCLONAL
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rabbit
Ig
RUO
Store undiluted at -20°C.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.