Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14121

Catalog No. PA14121


Add to cart

Description

Description

PA1-4121 detects neuronal Nitric Oxide Synthase in human samples. PA1-4121 has been successfully used in Western blot procedures. The PA1-4121 immunogen is a synthetic peptide corresponding to residues P(19) R K G P P L P N G D T E C(31) of nNOS.

Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) & endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains. eNOS and bNOS share ~50% sequence homology, and their activity depends on binding to the calcium/calmodulin complex. Both constitutive isoforms respond immediately to increased levels of calcium to produce low levels of NO over a short period of time. bNOS has been found in neurons, peripheral nerve cells, macula densa & pancreatic islet cells.
Specifications

Specifications

nNOS
Polyclonal
Unconjugated
NOS1
Liquid
NOS1
Synthetic peptide corresponding to residues P(19) R K G P P L P N G D T E C(31) of nNOS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P29475
Neuronal Nitric Oxide Synthase, NOS Type 1, Brain NOS, bNOS1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4842
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit