Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOB1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524749
Description
This antibody is predicted to react with bovine, mouse, non-human primate and rat based on sequence homology.
NOB1 may play a role in mRNA degradation.Specifications
NOB1 | |
Polyclonal | |
NOB1 | |
ART-4, MST158, NOB1P, PSMD8BP1, PSMD8 binding protein 1, RNA-binding protein NOB1, adenocarcinoma antigen recognized by T lymphocytes 4, nin one binding protein, phosphorylation regulatory protein HP-10, protein ART-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28987 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9ULX3 | |
NOB1 | |
KLH conjugated synthetic peptide between 383-412 amino acids from the C-terminal region of human NOB1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction