Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nociceptin Rabbit anti-Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA127972

Catalog No. PIPA127972


Add to cart

Description

Description

PA1-27972 detects Nociceptin from rat, non-human primate samples. PA1-27972 has been successfully used in immunohistochemistry (frozen tissue) applications. The PA1-27972 immunogen is: Synthetic peptide: FGGFTGARKSARKLANQ, corresponding to amino acids 122-138 of Rat Orphanin FQ. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Nociceptin is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. May be involved in neuronal differentiation and development.
Specifications

Specifications

Nociceptin
Polyclonal
whole serum with 0.05% sodium azide
Q62923
Orphanin FQ, PPNOC
Rabbit
IgG
RUO
Antibody
Polyclonal
Non-human Primate, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Pnoc
Liquid
PNOC
synthetic peptide FGGFTGARKSARKLANQ, corresponding to amino acids 122-138 of Rat Orphanin FQ.
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
25516
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit