Learn More
Nociceptin Rabbit anti-Guinea Pig, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118311
Catalog No. 01-670-564
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18311 detects Nociceptin from rat and Guinea Pig samples. PA1-18311 has been successfully used in immunohistochemistry and ELISA procedures. The PA1-18311 immunogen is a synthetic peptide corresponding to residues C-T(139) G A R K S A R K L A N Q(151) of rat Nociceptin peptide, conjugated to diphtheria toxoid. After reconstitution, store at -20C or 4C with an appropriate antibacterial agent. Reconstitute with 100 μL of distilled water. Glycerol (1:1) may be added for additional stability.
Nociceptin is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. May be involved in neuronal differentiation and development.
Specifications
Specifications
Nociceptin
Polyclonal
whole serum with no preservative
Q62923
Orphanin FQ, PPNOC
Rabbit
IgG
RUO
Antibody
Polyclonal
Guinea Pig, Rat
ELISA, Immunohistochemistry
Unconjugated
Pnoc
Lyophilized
PNOC
Synthetic peptide corresponding to residues C-T(139) G A R K S A R K L A N Q(151) of rat Nociceptin peptide.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100735883, 25516