Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOLA1, Polyclonal, Invitrogen™

Catalog No. PIPA127677
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127677 50 μg
1 options

Catalog No. PIPA127677

Supplier: Thermo Scientific PA127677

Chicken Polyclonal Antibody

Required for ribosome biogenesis and telomere maintenance. Part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex. which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequent

PA1-27677 detects NOLA1 from human and mouse samples.PA1-27677 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27720 immunogen is a synthetic peptide corresponding to human H/ACA ribonucleoprotein complex subunit 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOLA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene GAR1
Gene Accession No. Q9NY12, Q9CY66
Gene Alias NOLA1
Gene Symbols GAR1
Host Species Chicken
Immunogen Synthetic peptide corresponding to human H/ACA ribonucleoprotein complex subunit 1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54433
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.