Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH9 Rabbit anti-Canine, Human, Rat, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124943

Catalog No. PA124943


Add to cart

Description

Description

PA1-24943 detects non-muscle Myosin IIA from human, rat, Xenopus laevis and canine samples. PA1-24943 has been successfully used in Western blot procedures. The PA1-24943 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 1950-1961 of human non-muscle Myosin IIA. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

MYH9
Polyclonal
Unconjugated
MYH9
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100911597, 398083, 4627, 481280
Immunocytochemistry, Immunofluorescence, Western Blot
10.2 mg/mL
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
Q258K2, P35579, Q62812
MYH9, myh9.L
Synthetic peptide, conjugated to KLH, corresponding to residues 1950-1961 of human non-muscle Myosin IIA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human, Rat, Xenopus laevis
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit