Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH10 Rabbit anti-Canine, Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124944

Catalog No. PA124944


Add to cart

Description

Description

PA1-24944 detects non-muscle Myosin IIB from human, rat, and canine samples. PA1-24944 has been successfully used in Western blot procedures. The PA1-24944 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 1965-1976 of human non-muscle Myosin IIB. Store at 4°C short term. For extended storage, aliquot and store at -20°C, avoiding freeze/thaw cycles.

Specifications

Specifications

MYH10
Polyclonal
Unconjugated
MYH10
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4628, 479497, 79433
Western Blot
1 mg/mL
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
P35580, Q9JLT0
MYH10
Synthetic peptide, conjugated to KLH, corresponding to residues 1965-1976 of human non-muscle Myosin IIB.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit