Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP2 Mouse anti-Yeast, Clone: 22G1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA110027

Catalog No. 01-670-284


Add to cart

Description

Description

For Western blot using yeast samples, use a dilution of 1:2000 for cell lysates and 1:10,000 for nuclear fractions.

Nop2 is the yeast homologue of human p120 nucleolar proliferation antigen. This protein is localized in the nucleus.
Specifications

Specifications

Nop2
Monoclonal
Unconjugated
NOP2
MGC117384, MGC149287, MGC149288, NOL1, NOP120, NSUN1, p120, nop2p
Mouse
IgG3
RUO
Antibody
Monoclonal
Yeast
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
22G1
tissue culture supernatant with no preservative
Liquid
NOP2
Yeast nuclear preparation.
500 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
855664
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit