Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nop5/Nop6 Mouse anti-Yeast, Clone: 37C12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA110028

Catalog No. 01-670-285



Description

Description

Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.
Specifications

Specifications

Nop5/Nop6
Monoclonal
Unconjugated
NOP58
NOP58, NOP6, YOR310C, NOP58, Nucleolar protein 58, Nucleolar protein 5, Nucleolar protein 6
Mouse
IgG3
RUO
Antibody
Monoclonal
Yeast
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
37C12
tissue culture supernatant with no preservative
Liquid
NOP58, NOP6
Yeast nuclear preparation.
500 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
851313, 854487
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit