Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOR-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124536
Description
PA1-24536 detects NOR-1 in Human samples. PA1-24536 has been successfully used in Immunohistochemistry (paraffin) procedures. PA1-24536 immunogen corresponds to Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 65/75%. Rat: 65/75%.
NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3′ variant.Specifications
NOR-1 | |
Polyclonal | |
Unconjugated | |
Q92570 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8013 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
NR4A3 | |
NR4A3 | |
Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 65/75%. Rat: 65/75%. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction