Learn More
nNOS Guinea Pig anti-Human, Rat, Polyclonal, Invitrogen™
Guinea Pig Polyclonal Antibody
Manufacturer:  Invitrogen PA136024
Catalog No. 01-672-214
Add to cart
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins.
Specifications
Specifications
nNOS
Polyclonal
whole serum with no preservative
P29475, P29476
NOS
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry (Frozen)
Unconjugated
NOS1
Lyophilized
NOS1
Synthetic peptide from the C-terminal of rat cerebellar NO-synthase, conjugated to BSA.
50 μL
4° C
Primary
24598, 4842