Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Catalog No. 01672105
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
01-672-105 50 μL
1 options

Catalog No. 01-672-105

Supplier: Thermo Scientific PA136152

Rabbit Polyclonal Antibody

This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins.

Reconstitute with 50-100 ul of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen nNOS
Applications Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Gene NOS1
Gene Accession No. P29475, P29476
Gene Alias IHPS1, NOS, nNOS
Gene Symbols NOS1
Host Species Rabbit
Immunogen Synthetic fragment of rat cerebellar NO-synthase.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24598, 4842
Target Species Human, Rat
Content And Storage 4° C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.