Learn More
nNOS Goat anti-Guinea Pig, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen OSN00003W
Catalog No. PIOSN00003W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 150 µL of distilled water.
enzyme that catalyzes the production of nitric oxide.
Specifications
Specifications
nNOS
Polyclonal
whole serum with no preservative
P29475, Q9Z0J4, O19132, P29476
BNOS, NOS type I, Neuronal NOS, N-NOS, nNOS, Constitutive NOS, NC-NOS, Nitric oxide synthase
Goat
IgG
RUO
Antibody
Polyclonal
Guinea Pig, Human, Mouse, Rabbit, Rat
Immunohistochemistry, Western Blot
Unconjugated
NOS1
Lyophilized
NOS1
A synthetic pentamer-peptide from the c-terminal region of human NOS1
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100009243, 100729682, 18125, 24598, 4842