Learn More
NOS3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human NOS3 peptide using ARM Technology.
Supplier: Abnova Corporation H00004846K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. Nitric oxide is synthesized from L-arginine by nitric oxide synthases. Variations in this gene are associated with susceptibility to coronary spasm. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
NOS3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
NOS3 | |
NOS3 | |
A synthetic peptide of human NOS3 | |
RUO | |
4846 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
nitric oxide synthase 3 (endothelial cell) | |
NOS3 | |
ECNOS, eNOS | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.