Learn More
Notch 1 Syrian Hamster anti-Mouse, Rat, Clone: 8G10, MilliporeSigma™
Syrian Hamster Monoclonal Antibody
Manufacturer:  MilliporeSigma 05557
Catalog No. 05-557-MI
Add to cart
Description
Description
Detect Notch 1 with Anti-Notch 1 Antibody, clone 8G10 (Syrian Hamster Monoclonal Antibody), that has been shown to work in flow cytometry, immunocytochemistry and western blotting.
Specifications
Specifications
Notch 1
8G10
Cell Culture supernatant
Supernatant
Syrian Hamster
IgG
200μL
Primary
Murine, Rat
Flow Cytometry, Immunocytochemistry, Western Blot
Unconjugated
NM_017617
NOTCH1
Residues 1299-1492 of murine notch 1
Unpurified
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal