Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Notch-1 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems FAB5317P025
This item is not returnable.
View return policy
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human samples. It is validated for Flow Cytometry.Specifications
Notch-1 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4851 | |
Human | |
Purified |
Flow Cytometry | |
527425 | |
Flow Cytometry 10 uL/10^6 cells | |
P46531 | |
NOTCH1 | |
Mouse myeloma cell line NS0-derived recombinant human Notch-1 Ala19-Gln526 Accession # P46531 | |
25 Tests | |
Primary | |
Detects human Notch-1. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction