Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | Notch-2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227347SS
|
Novus Biologicals
NBP227347SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP227347
|
Novus Biologicals
NBP227347 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
Notch-2 Polyclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot.Specifications
Notch-2 | |
Western Blot 2-4ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2 | |
NOTCH2 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in pig and horse, and 88% homologous in mouse, rat, dog and bovine. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Signal Transduction, Stem Cell Signaling Pathway | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
4853 | |
A portion of amino acids 1550-1600 of humanNOTCH2 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title