Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch2 Mouse anti-Human, PE, Clone: MHN2-25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563519

Catalog No. BDB563519


Add to cart

Description

Description

The MHN2-25 monoclonal antibody specifically binds to an extracellular domain of human Notch2. Notch2 is a type 1 transmembrane glycoprotein receptor and member of the Notch family that includes Notch1-Notch4. Notch2 is cleaved in the Golgi and presents as a cell surface heterodimeric receptor. The Notch2 receptor can bind to several membrane-bound ligands including Jagged1, Jagged2, Delta1 and Delta4. Upon ligand binding, Notch2 undergoes proteolytic cleavage that results in the release of the Notch intracellular domain, NICD. NICD translocates to the nucleus where it forms a transcriptional activator complex with various transcriptional factors. These multimeric complexes either positively or negatively regulate the expression of multiple genes including those that orchestrate many facets of embryonic development and the subsequent functioning of multiple organ systems such as the hematopoietic, immune, cardiovascular, hepatic and renal systems. Notch2 is expressed by cells of the lung and brain, as well as cells of the B lineage, thymocytes, and T cells. Abnormalities in Notch2 expression may play a role in the development of B cell lymphomas.

Specifications

Specifications

Notch2
MHN2-25
NOTC2; AGS2; hN2; HJCYS; Neurogenic locus notch homolog protein 2
Affinity Purified
Human Notch2 Recombinant Protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG2a, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit