Learn More
Notch4 Mouse anti-Human, Alexa Fluor 647, Clone: MHN4-2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563904
Catalog No. BDB563904
Add to cart
Description
Description
The MHN4-2 monoclonal antibody specifically binds to an extracellular domain of Neurogenic locus notch homolog protein 4, also known as, Notch4 (or Notch 4 and Notch-4). Notch4 is a type 1 transmembrane glycoprotein that belongs to the Notch Receptor Family comprised of Notch1 through Notch4. Notch4 serves as a receptor for several ligands including Delta1, Jagged1, and Jagged2 that can be expressed on the same or different cells. Notch4 plays important roles in embryonic and postnatal development and cell fate determination by regulating cellular differentiation, proliferation and apoptosis. After ligand binding, the Notch4 receptor is cleaved in its extracellular and transmembrane domains. The cleaved notch intracellular domain (NICD) dissociates from the membrane and translocates to the nucleus where it can act as a transcriptional activator of downstream target genes involved with cellular differentiation and other responses. Notch4 is expressed at high levels in the heart and by endothelial cells and by certain CD34-positive and CD34-negative human bone marrow cell subsets. It is over-expressed by cells in certain breast, colon, and lung cancers.
Specifications
Specifications
Notch4
MHN4-2
Notch-4; Neurogenic locus notch homolog protein 4; hNotch4; NOTC4; INT3
Affinity Purified
Human Notch4 Transfected Cell Line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human