Learn More
Notch4 Mouse anti-Human, Brilliant Violet 421, Clone: MHN4-2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563905
Catalog No. BDB563905
Add to cart
Description
Description
The MHN4-2 monoclonal antibody specifically binds to an extracellular domain of Neurogenic locus notch homolog protein 4, also known as, Notch4 (or Notch 4 and Notch-4). Notch4 is a type 1 transmembrane glycoprotein that belongs to the Notch Receptor Family comprised of Notch1 through Notch4. Notch4 serves as a receptor for several ligands including Delta1, Jagged1, and Jagged2 that can be expressed on the same or different cells. Notch4 plays important roles in embryonic and postnatal development and cell fate determination by regulating cellular differentiation, proliferation and apoptosis. After ligand binding, the Notch4 receptor is cleaved in its extracellular and transmembrane domains. The cleaved notch intracellular domain (NICD) dissociates from the membrane and translocates to the nucleus where it can act as a transcriptional activator of downstream target genes involved with cellular differentiation and other responses. Notch4 is expressed at high levels in the heart and by endothelial cells and by certain CD34-positive and CD34-negative human bone marrow cell subsets. It is over-expressed by cells in certain breast, colon, and lung cancers.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifications
Specifications
Notch4
MHN4-2
Notch-4; Neurogenic locus notch homolog protein 4; hNotch4; NOTC4; INT3
Affinity Purified
Human Notch4 Transfected Cell Line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human