Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NOX1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP219568

Catalog No. NBP219568

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

NOX1
0.76mg/mL
0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7).
Affinity Purified
Recombinant fragment corresponding to a region within amino acids 201 and 456 of Mox1 (Uniprot ID No. Q9Y5S8)
RUO
Polyclonal
Species reactivity based on sequence identity: Mouse (86%), Rat (83%), Bovine (87%).
Western Blot
Unconjugated
Q9Y5S8
Rabbit
0.1mL
Primary
27035
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only