Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOXO1 Rabbit anti-Human, Chimpanzee, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177899

Catalog No. NBP177899


Add to cart

Description

Description

NOXO1 Polyclonal antibody specifically detects NOXO1 in Human, Chimpanzee samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

NOXO1
1 mg/ml
Western Blot 1:1000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Q8NFA2
MGC20258, NADPH oxidase organizer 1, Nox organizer 1, Nox-organizing protein 1, P41NOX, P41NOXA, P41NOXB, P41NOXC, regulatory protein P41NOX, SH3 and PX domain-containing protein 5, SH3PXD5NADPH oxidase regulatory protein, SNX28
Rabbit
IgG
0.1 mg
Apoptosis, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary
124056
Chimpanzee, Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
NOXO1
NOXO1 affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 238-252 of human NOXO1 protein.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
A BLAST analysis was used to suggest cross reactivity with NOXO1 protein from human and chimpanzee (100% homology). Expect reactivity with alpha, delta and gamma isoforms of NOXO1. Also expect partial reactivity against NOXO1 homologues from dog and guinea pig (87%), as well as rat (75%) and mouse (68%). Reactivity against homologues from other sources is not known.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only