Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1L5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526748

Catalog No. PIPA526748

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

Specifications

Specifications

NAP1L5
Polyclonal
PBS with 0.09% sodium azide
Q96NT1, Q9JJF0
DRLM, down-regulated in liver maligcy
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
266812, 58243
Western Blot
Unconjugated
NAP1L5
Liquid
NAP1L5
KLH conjugated synthetic peptide between 82-111 amino acids from the central region of human NP1L5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit