Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP50, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131827

Catalog No. PIPA131827

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.

PA1-31827 detects Npap60 + Nup50 from human samples.PA1-31827 has been successfully used in Western blot applications.The PA1-31827 immunogen is: Peptide with sequence ELHKILLEKKDA, from C Terminus of the protein sequence according to NP_009103; NP_705931; NP_710151.
Specifications

Specifications

NUP50
Polyclonal
Unconjugated
NUP50
Purified
NUP50
Peptide with sequence ELHKILLEKKDA, from C Terminus of the protein sequence according to NP_009103; NP_705931; NP_710151.
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine
Immunohistochemistry (Paraffin), Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9UKX7, Q9JIH2
CTA-268H5.7, MGC39961, NPAP60, NPAP60L
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
10762
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit