Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPAS1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124403

Catalog No. PIPA124403


Add to cart

Description

Description

PA1-24403 detects NPAS1 in Human samples. PA1-24403 has been successfully used in Western Blot procedures. PA1-24403 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human NPAS1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.
Specifications

Specifications

NPAS1
Polyclonal
Unconjugated
NPAS1
Liquid
NPAS1
Synthetic peptide region of human NPAS1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q99742
MOP5, PASD5, bHLHe11
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4861
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit