Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPAS2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528298
Description
The protein encoded by this gene is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. A similar mouse protein may play a regulatory role in the acquisition of specific types of memory. It also may function as a part of a molecular clock operative in the mammalian forebrain.
PA5-28298 targets NPAS2 in IF, IHC (P), and WB applications and shows reactivity with Human samples.The PA5-28298 immunogen is recombinant fragment corresponding to a region within amino acids 549 and 816 of Human NPAS2.Specifications
NPAS2 | |
Polyclonal | |
Unconjugated | |
Q99743 | |
NPAS2 | |
Recombinant fragment corresponding to a region within amino acids 549 and 816 of Human NPAS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3mg/mL | |
NPAS2 | |
MOP4, PASD4, bHLHe9, PAS domain-containing protein 4, basic-helix-loop-helix-PAS protein MOP4, class E basic helix-loop-helix protein 9, member of PAS protein 4, member of PAS superfamily 4, neuronal PAS domain-containing protein 2, neuronal PAS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4862 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction