Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528955

Catalog No. PIPA528955


Add to cart

Description

Description

PA5-28955 targets NPC2 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28955 immunogen is recombit fragment corresponding to a region within amino acids 1 and 151 of Human NPC2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein containing a lipid recognition domain. The encoded protein may function in regulating the transport of cholesterol through the late endosomal/lysosomal system. Mutations in this gene have been associated with Niemann-Pick disease, type C2 and frontal lobe atrophy.
Specifications

Specifications

NPC2
Polyclonal
Unconjugated
NPC2
Liquid
NPC2
Recombit fragment corresponding to a region within amino acids 1 and 151 of Human NPC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.69 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P61916
EDDM1, HE1, epididymal protein 1, epididymal secretory protein E1, human epididymis-specific protein 1, niemann-Pick disease type C2 protein, tissue-specific secretory protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10577
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit