Learn More
anti-NPIPL2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232740
Catalog No. NBP232740
Add to cart
Description
Description
NPIPL2 Polyclonal antibody specifically detects NPIPL2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
NPIPL2
0.5 mg/mL
Western Blot 1:1000
A6NHN6
A-761H5.4, LOC440348, nuclear pore complex interacting protein-like 2, Nuclear Pore Complex-Interacting Protein-Like 2
Rabbit
IgG
Protein A purified
RUO
Primary
440348
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
NPIPL2
This NPIPL2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 300-327 amino acids from the C-terminal region of human NPIPL2.
51.267 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human