Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPSR1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524177

Catalog No. PIPA524177


Add to cart

Description

Description

NPSR1 is a member of the G protein-coupled receptor 1 family and encodes a plasma membrane protein. Increased expression of this gene in ciliated cells of the respiratory epithelium and in bronchial smooth muscle cells is associated with asthma.
Specifications

Specifications

NPSR1
Polyclonal
PBS with 0.09% sodium azide
Q6W5P4, Q8BZP8
ASRT2, GPR154, GPRA, NPSR, PGR14, VRR1, G protein-coupled receptor 154, G protein-coupled receptor for asthma susceptibility, G-protein coupled receptor 154, G-protein coupled receptor PGR14, G-protein coupled receptor for asthma susceptibility, neuropeptide S receptor, vasopressin receptor-related r
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
319239, 387129
Western Blot
Unconjugated
NPSR1
Liquid
NPSR1
KLH conjugated synthetic peptide between 327-356 amino acids from the C-terminal region of human NPSR1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit